Product Details

SNP ID
rs114973583
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:161984250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTGTGCCGATCCAGCTGTACG[A/G]GAGCTTGTAGGAATTGCTCCAGCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFML2B PubMed Links

Gene Details

Gene
OLFML2B
Gene Name
olfactomedin like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297713.1 2719 Missense Mutation CCG,TCG P561S NP_001284642.1
NM_015441.2 2719 Missense Mutation CCG,TCG P560S NP_056256.1
XM_005245075.2 2719 Missense Mutation CCG,TCG P562S XP_005245132.1
XM_011509398.2 2719 Missense Mutation CCG,TCG P320S XP_011507700.1
XM_017000967.1 2719 Intron XP_016856456.1

View Full Product Details