Product Details

SNP ID
rs115074338
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:247122732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGAGTTTTGCAGTAACACCCA[C/T]TGAAGTGCACAAGGCGATGACCTTT
Phenotype
MIM: 194631
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF124 PubMed Links

Gene Details

Gene
ZNF124
Gene Name
zinc finger protein 124
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243740.2 1498 UTR 3 NP_001230669.1
NM_001297567.1 1498 Intron NP_001284496.1
NM_001297568.1 1498 Intron NP_001284497.1
NM_001297569.1 1498 Intron NP_001284498.1
NM_003431.3 1498 Intron NP_003422.2
XM_005273257.4 1498 Intron XP_005273314.1
XM_006711813.3 1498 Intron XP_006711876.1
XM_011544272.2 1498 Intron XP_011542574.1

View Full Product Details