Product Details

SNP ID
rs115133140
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:227921692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCACGGTGGCACCTACGGCCAGC[A/G]GTGCCCGGGGAGAAGCGGCCAGCCA
Phenotype
MIM: 602863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT9A PubMed Links

Gene Details

Gene
WNT9A
Gene Name
Wnt family member 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003395.2 1495 Silent Mutation ACC,ACT T308T NP_003386.1
XM_011544271.2 1495 Silent Mutation ACC,ACT T238T XP_011542573.1

View Full Product Details