Product Details

SNP ID
rs115176407
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:152776859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGTCCTGGAAGATTCCTCCGAC[C/G]TAGAATCCAGAAATCTGCCCTCTCA
Phenotype
MIM: 612607 MIM: 612608
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LCE1E PubMed Links

Gene Details

Gene
LCE1E
Gene Name
late cornified envelope 1E
There are no transcripts associated with this gene.

Gene
LCE1F
Gene Name
late cornified envelope 1F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178354.2 488 UTR 3 NP_848131.1

View Full Product Details