Product Details

SNP ID
rs115296365
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:215577112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAGCATCATTTCATCTGGATAA[C/T]ATAGCCCTTCCCAAGGCTAGTCTTT
Phenotype
MIM: 613272
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD3 PubMed Links

Gene Details

Gene
KCTD3
Gene Name
potassium channel tetramerization domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319294.1 Intron NP_001306223.1
NM_001319295.1 Intron NP_001306224.1
NM_016121.4 Intron NP_057205.2
XM_005273158.1 Intron XP_005273215.1

View Full Product Details