Product Details

SNP ID
rs115304948
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:226878881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTGAGAAAATGCAGCAACTCTT[C/T]GTTGTGGTGCCCCTCTTTGAGTAAG
Phenotype
MIM: 600759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSEN2 PubMed Links

Gene Details

Gene
PSEN2
Gene Name
presenilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000447.2 Intron NP_000438.2
NM_012486.2 Intron NP_036618.2
XM_005273199.3 Intron XP_005273256.1
XM_017001835.1 Intron XP_016857324.1
XM_017001836.1 Intron XP_016857325.1

View Full Product Details