Product Details

SNP ID
rs115343456
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:28593067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAAGTGTCGAGGAGATGTGGAA[A/G]GGAGAGAGGATCAGCAGGAGTTTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB42 PubMed Links

Gene Details

Gene
RAB42
Gene Name
RAB42, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193532.1 Intron NP_001180461.1
NM_152304.1 Intron NP_689517.1
XM_017000226.1 Intron XP_016855715.1

View Full Product Details