Product Details

SNP ID
rs115435412
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:20501113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGTACTGCATGCCTTGTTTGGG[C/T]GGCTGCAGGCGGACAGAGTTGTTGT
Phenotype
MIM: 612037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUL1 PubMed Links

Gene Details

Gene
MUL1
Gene Name
mitochondrial E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024544.2 1476 Silent Mutation CCA,CCG P212P NP_078820.2
XM_011542137.2 1476 Silent Mutation CCA,CCG P173P XP_011540439.1

View Full Product Details