Product Details

SNP ID
rs115694135
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:200554422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGGAAAGATGTCTAAGGTTCC[C/T]TAAAATATAAAATTCTGATTATAAA
Phenotype
MIM: 611279
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF14 PubMed Links

Gene Details

Gene
KIF14
Gene Name
kinesin family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305792.1 Intron NP_001292721.1
NM_014875.2 Intron NP_055690.1
XM_011510231.2 Intron XP_011508533.1
XM_011510232.2 Intron XP_011508534.1
XM_011510233.2 Intron XP_011508535.1
XM_011510235.2 Intron XP_011508537.1
XM_011510236.1 Intron XP_011508538.1
XM_017003005.1 Intron XP_016858494.1
XM_017003006.1 Intron XP_016858495.1
XM_017003007.1 Intron XP_016858496.1

View Full Product Details