Product Details

SNP ID
rs115904929
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:155085064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTTTGCTCTGGGGTTTCTTCT[C/G]TCTGAGCCGCTTCCTCTTCCCCACA
Phenotype
MIM: 601381
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EFNA3 PubMed Links

Gene Details

Gene
EFNA3
Gene Name
ephrin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004952.4 Intron NP_004943.1

View Full Product Details