Product Details

SNP ID
rs115917975
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:153607388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGACTGAGACACTCACAAAGGGA[A/C]CCCAGTTCAGCAAGGGTCAGAGGAA
Phenotype
MIM: 607986
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
S100A14 PubMed Links

Gene Details

Gene
S100A14
Gene Name
S100 calcium binding protein A14
There are no transcripts associated with this gene.

Gene
S100A16
Gene Name
S100 calcium binding protein A16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317007.1 701 UTR 3 NP_001303936.1
NM_001317008.1 701 UTR 3 NP_001303937.1
NM_080388.2 701 UTR 3 NP_525127.1

View Full Product Details