Product Details

SNP ID
rs115976195
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:34757325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGTAGCAGCTGACGCGTGGG[C/T]CCACCATGAACTGGAGTATCTTTGA
Phenotype
MIM: 605425 MIM: 604493
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJB4 PubMed Links

Gene Details

Gene
GJB4
Gene Name
gap junction protein beta 4
There are no transcripts associated with this gene.

Gene
GJB5
Gene Name
gap junction protein beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005268.3 435 UTR 5 NP_005259.1
XM_005270751.3 435 UTR 5 XP_005270808.1

View Full Product Details