Product Details

SNP ID
rs116221847
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:109214455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCGGGGCGGGAGGTTGTGGGG[G/T]CTACGCGGCTTTCCTAACACGAATC
Phenotype
MIM: 611298 MIM: 607529
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1324 PubMed Links

Gene Details

Gene
KIAA1324
Gene Name
KIAA1324
There are no transcripts associated with this gene.

Gene
SARS
Gene Name
seryl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006513.3 Intron NP_006504.2
XM_006710813.2 Intron XP_006710876.1

View Full Product Details