Product Details

SNP ID
rs116299558
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:47218601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTAGCCAGGGCCTGGTCTCTGA[C/T]TGGATTTCCCATACCCTAGAAGTGA
Phenotype
MIM: 187040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAL1 PubMed Links

Gene Details

Gene
TAL1
Gene Name
TAL bHLH transcription factor 1, erythroid differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287347.2 2441 UTR 3 NP_001274276.1
NM_001290403.1 2441 UTR 3 NP_001277332.1
NM_001290404.1 2441 UTR 3 NP_001277333.1
NM_001290405.1 2441 UTR 3 NP_001277334.1
NM_001290406.1 2441 UTR 3 NP_001277335.1
NM_003189.5 2441 UTR 3 NP_003180.1
XM_005271160.4 2441 UTR 3 XP_005271217.1
XM_017002187.1 2441 UTR 3 XP_016857676.1
XM_017002188.1 2441 UTR 3 XP_016857677.1
XM_017002189.1 2441 UTR 3 XP_016857678.1
XM_017002190.1 2441 UTR 3 XP_016857679.1
XM_017002191.1 2441 UTR 3 XP_016857680.1
XM_017002192.1 2441 Intron XP_016857681.1
XM_017002193.1 2441 UTR 3 XP_016857682.1

View Full Product Details