Product Details

SNP ID
rs116527678
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:145921126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAAAAGTAATATGCATCTCCAG[G/C]GCCACTCAAATTTTGCTAAGGACAG
Phenotype
MIM: 612875 MIM: 603867 MIM: 605313
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
GNRHR2 PubMed Links

Gene Details

Gene
GNRHR2
Gene Name
gonadotropin releasing hormone receptor 2 (pseudogene)
There are no transcripts associated with this gene.

Gene
PEX11B
Gene Name
peroxisomal biogenesis factor 11 beta
There are no transcripts associated with this gene.

Gene
RBM8A
Gene Name
RNA binding motif protein 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005105.4 Intron NP_005096.1

View Full Product Details