Product Details

SNP ID
rs116558689
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:78650528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGTTACAAAATATAACTCCCC[A/G]ACTAATTTCCAGATAGATGGAAGAA
Phenotype
MIM: 610468 MIM: 613975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI44 PubMed Links

Gene Details

Gene
IFI44
Gene Name
interferon induced protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006417.4 1695 Silent Mutation CCA,CCG P111P NP_006408.3
XM_005270380.3 1695 Silent Mutation CCA,CCG P111P XP_005270437.1
XM_011540516.2 1695 Silent Mutation CCA,CCG P111P XP_011538818.1
XM_017000083.1 1695 Silent Mutation CCA,CCG P111P XP_016855572.1
Gene
IFI44L
Gene Name
interferon induced protein 44 like
There are no transcripts associated with this gene.

View Full Product Details