Product Details

SNP ID
rs116602746
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:6525591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTGAGGAATCTCTGTTTTAAT[A/G]GCACACAAACTGTTCTCTGCTGTTT
Phenotype
MIM: 611101
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOL9 PubMed Links

Gene Details

Gene
NOL9
Gene Name
nucleolar protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024654.4 1394 UTR 3 NP_078930.3
XM_005263493.3 1394 UTR 3 XP_005263550.1
XM_011542147.2 1394 UTR 3 XP_011540449.1
XM_017002337.1 1394 Intron XP_016857826.1
XM_017002338.1 1394 UTR 3 XP_016857827.1
XM_017002339.1 1394 UTR 3 XP_016857828.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

View Full Product Details