Product Details

SNP ID
rs116631538
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:205303773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGGGCGAGTTCATCCAGGGAGC[C/T]GAAGGTGGTGGGGGCCGCGAGCTCC
Phenotype
MIM: 608131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUAK2 PubMed Links

Gene Details

Gene
NUAK2
Gene Name
NUAK family kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030952.2 984 Missense Mutation AGC,GGC S566G NP_112214.2
XM_005245515.4 984 Missense Mutation AGC,GGC S275G XP_005245572.1

View Full Product Details