Product Details

SNP ID
rs116637066
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:23084128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGTGGTGTACATTTTGGGAAGC[A/G]CACTACATAGGATCAAGGGCTTAGG
Phenotype
MIM: 609132 MIM: 601422
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM1A PubMed Links

Gene Details

Gene
KDM1A
Gene Name
lysine demethylase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009999.2 8382 Intron NP_001009999.1
NM_015013.3 8382 Intron NP_055828.2
XM_005245786.2 8382 Intron XP_005245843.1
XM_006710472.3 8382 Intron XP_006710535.1
XM_006710473.2 8382 Intron XP_006710536.1
XM_006710474.3 8382 Intron XP_006710537.1
XM_017000716.1 8382 Intron XP_016856205.1
XM_017000717.1 8382 Intron XP_016856206.1
XM_017000718.1 8382 Intron XP_016856207.1
XM_017000719.1 8382 Intron XP_016856208.1
XM_017000720.1 8382 Intron XP_016856209.1
Gene
LUZP1
Gene Name
leucine zipper protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142546.1 8382 UTR 3 NP_001136018.1
NM_033631.3 8382 UTR 3 NP_361013.3
XM_011542090.2 8382 UTR 3 XP_011540392.1
XM_011542091.2 8382 UTR 3 XP_011540393.1
XM_017002252.1 8382 UTR 3 XP_016857741.1

View Full Product Details