Product Details

SNP ID
rs116697924
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:173185595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAGCTGAGTGCGTATCTCAGCA[C/T]AGGCAGGAGGATGAGCATGTGTTGC
Phenotype
MIM: 603594
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF4 PubMed Links

Gene Details

Gene
TNFSF4
Gene Name
tumor necrosis factor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297562.1 2097 UTR 3 NP_001284491.1
NM_003326.4 2097 UTR 3 NP_003317.1
XM_011509964.2 2097 UTR 3 XP_011508266.2
XM_017002228.1 2097 UTR 3 XP_016857717.1
XM_017002229.1 2097 UTR 3 XP_016857718.1
XM_017002230.1 2097 UTR 3 XP_016857719.1

View Full Product Details