Product Details

SNP ID
rs116713004
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:62204061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTCTAGAAGAGTTTGGGCACAG[C/T]TAAGATAATAGTTGCTTCTAAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
L1TD1 PubMed Links

Gene Details

Gene
L1TD1
Gene Name
LINE1 type transposase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details