Product Details

SNP ID
rs117426382
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:215624757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCCAAAAAGAAGGGATATAGT[A/G]CAGAGTAATATATACATGATATACA
Phenotype
MIM: 613272 MIM: 608400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD3 PubMed Links

Gene Details

Gene
KCTD3
Gene Name
potassium channel tetramerization domain containing 3
There are no transcripts associated with this gene.

Gene
USH2A
Gene Name
usherin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007123.5 17020 Intron NP_009054.5
NM_206933.2 17020 UTR 3 NP_996816.2

View Full Product Details