Product Details

SNP ID
rs117489792
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27412630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAGAAGGAAGGAGAAGGTGAAG[A/C]AGCAGAGTCTGACTGTGGTGGTGGC
Phenotype
MIM: 605875
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WASF2 PubMed Links

Gene Details

Gene
WASF2
Gene Name
WAS protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201404.2 Intron NP_001188333.1
NM_006990.4 Intron NP_008921.1

View Full Product Details