Product Details

SNP ID
rs121908144
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:54999221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGACCTTCCGGATCGGCTTCA[C/T]TGTGCTGGGGCTTTTCCTGCTGGCC
Phenotype
MIM: 606412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSND PubMed Links

Gene Details

Gene
BSND
Gene Name
barttin CLCNK type accessory beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057176.2 278 Missense Mutation ACT,ATT T12I NP_476517.1
Gene
TMEM61
Gene Name
transmembrane protein 61
There are no transcripts associated with this gene.

View Full Product Details