Product Details

SNP ID
rs137902270
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:9105585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCTGAGGCCGATGAAGATGAG[C/T]GGGATGAGCACCAGCTTCTTGTCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR157 PubMed Links

Gene Details

Gene
GPR157
Gene Name
G protein-coupled receptor 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024980.4 746 Silent Mutation CCA,CCG P231P NP_079256.4
XM_005263496.4 746 Silent Mutation CCA,CCG P218P XP_005263553.1
XM_005263497.4 746 Intron XP_005263554.1

View Full Product Details