Product Details

SNP ID
rs138053747
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:205088478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAATGAAGTTAGATGAGCAAAAC[A/G]TAAGTATCTTTTCTCCAGCAACATA
Phenotype
MIM: 600697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBBP5 PubMed Links

Gene Details

Gene
RBBP5
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193272.1 2070 UTR 3 NP_001180201.1
NM_001193273.1 2070 UTR 3 NP_001180202.1
NM_005057.3 2070 UTR 3 NP_005048.2
XM_011509833.2 2070 UTR 3 XP_011508135.1
Gene
TMEM81
Gene Name
transmembrane protein 81
There are no transcripts associated with this gene.

View Full Product Details