Product Details

SNP ID
rs138096568
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:158420266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCCAGGACCAATGTACAGAGCA[A/T]GAAGATGACAATCTGACTAAAGTGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10K2 PubMed Links

Gene Details

Gene
OR10K2
Gene Name
olfactory receptor family 10 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004476.1 601 Missense Mutation ATG,TTG M201L NP_001004476.1

View Full Product Details