Product Details

SNP ID
rs138116431
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:46635824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTTCAGTTCTGCTCCAAGTCC[A/G]CTTTGCTCCAATTCAGCGATGACAG
Phenotype
MIM: 608917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042546.2 996 Silent Mutation AGC,AGT S268S NP_001036011.2
NM_001243728.1 996 Silent Mutation AGC,AGT S225S NP_001230657.1
NM_001256418.1 996 Silent Mutation AGC,AGT S162S NP_001243347.1
NM_022745.4 996 Silent Mutation AGC,AGT S336S NP_073582.3

View Full Product Details