Product Details

SNP ID
rs138242807
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:33518937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGATATGGGGTATATGTATTGC[A/G]TTTATAGTGTATGAATGTATATATA
Phenotype
MIM: 608398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSMD2 PubMed Links

Gene Details

Gene
CSMD2
Gene Name
CUB and Sushi multiple domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281956.1 11576 Intron NP_001268885.1
NM_052896.4 11576 Intron NP_443128.2
XM_017000185.1 11576 UTR 3 XP_016855674.1
XM_017000186.1 11576 Intron XP_016855675.1
XM_017000187.1 11576 Intron XP_016855676.1
XM_017000188.1 11576 UTR 3 XP_016855677.1
XM_017000189.1 11576 Intron XP_016855678.1
XM_017000190.1 11576 UTR 3 XP_016855679.1
XM_017000191.1 11576 Intron XP_016855680.1
XM_017000192.1 11576 Intron XP_016855681.1
XM_017000193.1 11576 Intron XP_016855682.1
XM_017000194.1 11576 Intron XP_016855683.1

View Full Product Details