Product Details

SNP ID
rs138247086
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:178483057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCTTTATTACTATCCCTATTC[C/T]CAACATTTCATGCTGTCTTTCATGA
Phenotype
MIM: 606136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00083 PubMed Links

Gene Details

Gene
LINC00083
Gene Name
long intergenic non-protein coding RNA 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017003086.1 Intron XP_016858575.1
Gene
RASAL2
Gene Name
RAS protein activator like 2
There are no transcripts associated with this gene.

View Full Product Details