Product Details

SNP ID
rs138253207
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:155131366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGAATGAGGACTACACCATACA[A/T]GTGCAGCTGAATGACTACGTGGACA
Phenotype
MIM: 605951 MIM: 191164 MIM: 613683
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DPM3 PubMed Links

Gene Details

Gene
DPM3
Gene Name
dolichyl-phosphate mannosyltransferase subunit 3
There are no transcripts associated with this gene.

Gene
EFNA1
Gene Name
ephrin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004428.2 225 Silent Mutation CAA,CAT Q40H NP_004419.2
NM_182685.1 225 Silent Mutation CAA,CAT Q40H NP_872626.1
Gene
SLC50A1
Gene Name
solute carrier family 50 member 1
There are no transcripts associated with this gene.

View Full Product Details