Product Details

SNP ID
rs138267967
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:74568205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACAAGTAATTTTATCAACATAG[A/T]ATTTGCAGAGAAAGAATTCACTTAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ERICH3 PubMed Links

Gene Details

Gene
ERICH3
Gene Name
glutamate rich 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002912.4 7065 UTR 3 NP_001002912.4
XM_017000274.1 7065 UTR 3 XP_016855763.1
XM_017000275.1 7065 Intron XP_016855764.1
XM_017000276.1 7065 UTR 3 XP_016855765.1
XM_017000277.1 7065 UTR 3 XP_016855766.1
XM_017000278.1 7065 UTR 3 XP_016855767.1
XM_017000279.1 7065 UTR 3 XP_016855768.1
XM_017000280.1 7065 UTR 3 XP_016855769.1
XM_017000281.1 7065 UTR 3 XP_016855770.1
XM_017000282.1 7065 UTR 3 XP_016855771.1
XM_017000283.1 7065 UTR 3 XP_016855772.1
XM_017000284.1 7065 UTR 3 XP_016855773.1
XM_017000285.1 7065 UTR 3 XP_016855774.1
Gene
ERICH3-AS1
Gene Name
ERICH3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details