Product Details

SNP ID
rs138630887
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:44806529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATGGAGCCCCGGCGCGAGGCC[A/G]GGGCTGGACCTGGACCTGCCGGTCG
Phenotype
MIM: 602913 MIM: 611713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136537.1 283 Intron NP_001130009.1
XM_006710386.3 283 Intron XP_006710449.1
XM_011540803.2 283 Intron XP_011539105.1
XM_011540805.2 283 Intron XP_011539107.1
XM_017000447.1 283 Intron XP_016855936.1
XM_017000448.1 283 Intron XP_016855937.1
XM_017000449.1 283 Intron XP_016855938.1
Gene
PLK3
Gene Name
polo like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004073.3 283 Intron NP_004064.2
Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 283 Missense Mutation NP_001013654.1
XM_006710614.3 283 Missense Mutation XP_006710677.1
XM_011541375.2 283 Missense Mutation XP_011539677.1
XM_011541376.2 283 Missense Mutation XP_011539678.1
XM_017001187.1 283 Missense Mutation XP_016856676.1

View Full Product Details