Product Details

SNP ID
rs138638208
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:34851984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACCCTGACCTGTCTATACCCAC[A/G]TCCACATAAACCTTGCTATCTAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM12 PubMed Links

Gene Details

Gene
SMIM12
Gene Name
small integral membrane protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164824.1 4164 UTR 3 NP_001158296.1
NM_001164825.1 4164 UTR 3 NP_001158297.1
NM_001320261.1 4164 UTR 3 NP_001307190.1
NM_138428.5 4164 UTR 3 NP_612437.3
XM_005270403.4 4164 Intron XP_005270460.1
XM_011540553.2 4164 Intron XP_011538855.1
XM_011540554.2 4164 Intron XP_011538856.2
XM_011540555.2 4164 Intron XP_011538857.1
XM_017000166.1 4164 Intron XP_016855655.1

View Full Product Details