Product Details

SNP ID
rs138654516
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:26023060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTGCCTCCTCCTCCTCCTCAG[A/C]CTGGACGCCCAGACTGGAGAGTGCA
Phenotype
MIM: 601738
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EXTL1 PubMed Links

Gene Details

Gene
EXTL1
Gene Name
exostosin like glycosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004455.2 1312 Missense Mutation AGA,AGC R138S NP_004446.2
XM_005245779.3 1312 Missense Mutation AGA,AGC R138S XP_005245836.1
XM_017000650.1 1312 Missense Mutation AGA,AGC R138S XP_016856139.1

View Full Product Details