Product Details

SNP ID
rs138746212
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:247606033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACAGCTGGCATCTATCTCCTGG[A/C]TCAGTGGTTTGGCTAGTTCCCTAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2G3 PubMed Links

Gene Details

Gene
OR2G3
Gene Name
olfactory receptor family 2 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001914.1 448 Missense Mutation ATC,CTC I150L NP_001001914.1

View Full Product Details