Product Details

SNP ID
rs139051375
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:161749990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGCTAACAGTGGACTCGGAGGA[A/G]CCCAGCTTCAAGGCGGGGCCTGGGG
Phenotype
MIM: 604835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP12 PubMed Links

Gene Details

Gene
DUSP12
Gene Name
dual specificity phosphatase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007240.2 223 Silent Mutation GAA,GAG E63E NP_009171.1
XM_005244862.2 223 UTR 5 XP_005244919.1

View Full Product Details