Product Details

SNP ID
rs139140747
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:23089052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGTACAGTCTTCCCCTTCTTCC[G/T]CCTGTGCCAATAAATAACAAAAGTG
Phenotype
MIM: 609132 MIM: 601422
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KDM1A PubMed Links

Gene Details

Gene
KDM1A
Gene Name
lysine demethylase 1A
There are no transcripts associated with this gene.

Gene
LUZP1
Gene Name
leucine zipper protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142546.1 3458 Missense Mutation NP_001136018.1
NM_033631.3 3458 Missense Mutation NP_361013.3
XM_011542090.2 3458 Missense Mutation XP_011540392.1
XM_011542091.2 3458 Missense Mutation XP_011540393.1
XM_017002252.1 3458 Missense Mutation XP_016857741.1

View Full Product Details