Product Details

SNP ID
rs139193605
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:46279032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCAAAGGCTCCACGGCCCCAGC[A/G]CTCCAGCCGCTTGGCGAACTCCAGA
Phenotype
MIM: 603615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC41 PubMed Links

Gene Details

Gene
LRRC41
Gene Name
leucine rich repeat containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006369.4 2316 Missense Mutation CGC,TGC R758C NP_006360.3
XM_017000074.1 2316 Intron XP_016855563.1
Gene
RAD54L
Gene Name
RAD54-like (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142548.1 2316 Intron NP_001136020.1
NM_003579.3 2316 Intron NP_003570.2
XM_006710975.3 2316 Intron XP_006711038.1
XM_011542299.2 2316 Intron XP_011540601.1
XM_011542300.2 2316 Intron XP_011540602.1

View Full Product Details