Product Details
- SNP ID
-
rs139206389
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:157092706 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTCTCCAGATTGGGGGAAGTAAG[G/T]CTTTCCTCCCCAGTTTTGAGTCTGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ETV3L
PubMed Links
Gene Details
- Gene
- ETV3L
- Gene Name
- ETS variant 3 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004341.2 |
1314 |
Missense Mutation |
AGA,AGC |
R343S |
NP_001004341.1 |
View Full Product Details