Product Details

SNP ID
rs139210753
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:45621315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCCTGCTGGCAAGGATGGCAC[A/C]GTTGCCCATGGGCCCGGGAGCAGGA
Phenotype
MIM: 603185
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC17 PubMed Links

Gene Details

Gene
CCDC17
Gene Name
coiled-coil domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114938.2 2331 Missense Mutation NP_001108410.2
NM_001190182.1 2331 Missense Mutation NP_001177111.1
XM_011540812.1 2331 Missense Mutation XP_011539114.1
XM_011540814.1 2331 Missense Mutation XP_011539116.1
XM_011540820.1 2331 Intron XP_011539122.1
XM_017000450.1 2331 Missense Mutation XP_016855939.1
XM_017000451.1 2331 Missense Mutation XP_016855940.1
XM_017000452.1 2331 Missense Mutation XP_016855941.1
XM_017000453.1 2331 Missense Mutation XP_016855942.1
XM_017000454.1 2331 Missense Mutation XP_016855943.1
XM_017000455.1 2331 Missense Mutation XP_016855944.1
XM_017000456.1 2331 Missense Mutation XP_016855945.1
XM_017000457.1 2331 Intron XP_016855946.1
XM_017000458.1 2331 Intron XP_016855947.1
Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
NASP
Gene Name
nuclear autoantigenic sperm protein
There are no transcripts associated with this gene.

View Full Product Details