Product Details

SNP ID
rs139218932
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:220061741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGATGAACTCAAATTGGACAC[A/G]CTAAATTTGAGATACCTGAGGGATG
Phenotype
MIM: 604053
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPNT1 PubMed Links

Gene Details

Gene
BPNT1
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286149.1 Intron NP_001273078.1
NM_001286150.1 Intron NP_001273079.1
NM_001286151.1 Intron NP_001273080.1
NM_006085.5 Intron NP_006076.4
XM_005272998.2 Intron XP_005273055.1
XM_005272999.4 Intron XP_005273056.1
XM_005273000.4 Intron XP_005273057.1
XM_005273004.2 Intron XP_005273061.1
XM_005273005.4 Intron XP_005273062.1
XM_006711114.3 Intron XP_006711177.1
XM_011509063.2 Intron XP_011507365.1
XM_011509064.2 Intron XP_011507366.1
XM_011509065.2 Intron XP_011507367.1
XM_017000043.1 Intron XP_016855532.1
XM_017000044.1 Intron XP_016855533.1
XM_017000045.1 Intron XP_016855534.1
XM_017000046.1 Intron XP_016855535.1

View Full Product Details