Product Details

SNP ID
rs139259107
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:161723536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTCTGCCCTCTCACAAGAAGAG[A/C]ATTGAGGTGCAGACACCAGGGGTGT
Phenotype
MIM: 606891 MIM: 609251
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FCRLA PubMed Links

Gene Details

Gene
FCRLA
Gene Name
Fc receptor like A
There are no transcripts associated with this gene.

Gene
FCRLB
Gene Name
Fc receptor like B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002901.3 242 Missense Mutation AGA,AGC R74S NP_001002901.1
NM_001288829.1 242 Missense Mutation AGA,AGC R74S NP_001275758.1
NM_001288830.1 242 Missense Mutation AGA,AGC R67S NP_001275759.1
NM_001288831.1 242 Missense Mutation AGA,AGC R74S NP_001275760.1
NM_001288832.1 242 Missense Mutation AGA,AGC R67S NP_001275761.1
NM_001320241.1 242 Missense Mutation AGA,AGC R74S NP_001307170.1

View Full Product Details