Product Details

SNP ID
rs139594365
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:161072841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGTCTGTGTTTCTATCTCCCT[C/T]ACCGTGGTCAGCGTGGAGTAACTGC
Phenotype
MIM: 614264 MIM: 609607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030916.2 1548 Silent Mutation GTA,GTG V451V NP_112178.2
XM_005245508.3 1548 Silent Mutation GTA,GTG V426V XP_005245565.1
XM_011510021.2 1548 Silent Mutation GTA,GTG V452V XP_011508323.1
XM_011510022.2 1548 Silent Mutation GTA,GTG V427V XP_011508324.1
XM_011510023.1 1548 UTR 3 XP_011508325.1

View Full Product Details