Product Details

SNP ID
rs139693961
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:161751713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGGCCATAATAACTGCTTTTCT[A/C]ATGAAGACTGACCAACTTCCCTTTG
Phenotype
MIM: 604835
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DUSP12 PubMed Links

Gene Details

Gene
DUSP12
Gene Name
dual specificity phosphatase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007240.2 424 Silent Mutation CTA,CTC L130L NP_009171.1
XM_005244862.2 424 UTR 5 XP_005244919.1

View Full Product Details