Product Details

SNP ID
rs139839658
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:109092985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCGTCTCAAAACCTGGCTGCTA[A/T]CAGAGAAGAAGGGTGTTTGGGGTGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SCARNA2 PubMed Links

Gene Details

Gene
SCARNA2
Gene Name
small Cajal body-specific RNA 2
There are no transcripts associated with this gene.

Gene
TMEM167B
Gene Name
transmembrane protein 167B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322248.1 198 Missense Mutation ACA,TCA T36S NP_001309177.1
NM_020141.3 198 Missense Mutation ACA,TCA T36S NP_064526.1

View Full Product Details