Product Details
- SNP ID
-
rs139857791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:74571257 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGTAGACTCTCCGGACTCAATT[C/G]CCTCTCTCCCTCCCGTGATAATCCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ERICH3
PubMed Links
Gene Details
- Gene
- ERICH3
- Gene Name
- glutamate rich 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001002912.4 |
4672 |
Missense Mutation |
CAA,GAA |
Q1485E |
NP_001002912.4 |
XM_017000274.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q1485E |
XP_016855763.1 |
XM_017000275.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q1483E |
XP_016855764.1 |
XM_017000276.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q803E |
XP_016855765.1 |
XM_017000277.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q789E |
XP_016855766.1 |
XM_017000278.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855767.1 |
XM_017000279.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855768.1 |
XM_017000280.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855769.1 |
XM_017000281.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855770.1 |
XM_017000282.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855771.1 |
XM_017000283.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855772.1 |
XM_017000284.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855773.1 |
XM_017000285.1 |
4672 |
Missense Mutation |
CAA,GAA |
Q743E |
XP_016855774.1 |
- Gene
- ERICH3-AS1
- Gene Name
- ERICH3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details