Product Details

SNP ID
rs139892328
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:235113852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTAAGACCTGCAGTAACTGCTG[C/T]GGCTGTCCACACACAGCAATTGCAT
Phenotype
MIM: 601848
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOMM20 PubMed Links

Gene Details

Gene
TOMM20
Gene Name
translocase of outer mitochondrial membrane 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014765.2 Intron NP_055580.1

View Full Product Details