Product Details

SNP ID
rs140091620
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:31301724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCAGTGGCCTAAGAGTAATAG[A/G]TTTAATTATTTCCATAATTCCTTTA
Phenotype
MIM: 607797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNRNP40 PubMed Links

Gene Details

Gene
SNRNP40
Gene Name
small nuclear ribonucleoprotein U5 subunit 40
There are no transcripts associated with this gene.

Gene
ZCCHC17
Gene Name
zinc finger CCHC-type containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282566.1 Intron NP_001269495.1
NM_001282567.1 Intron NP_001269496.1
NM_001282568.1 Intron NP_001269497.1
NM_001282569.1 Intron NP_001269498.1
NM_001282570.1 Intron NP_001269499.1
NM_001282571.1 Intron NP_001269500.1
NM_001282572.1 Intron NP_001269501.1
NM_001282573.1 Intron NP_001269502.1
NM_001282574.1 Intron NP_001269503.1
NM_016505.3 Intron NP_057589.2
XM_006710681.3 Intron XP_006710744.1
XM_011541567.2 Intron XP_011539869.1

View Full Product Details