Product Details

SNP ID
rs140241215
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:212836044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAGCTGACTTGATTGCTTCGTT[C/T]CACCAAGCAGTTGGATTCACGCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA45 PubMed Links

Gene Details

Gene
SPATA45
Gene Name
spermatogenesis associated 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024601.2 214 Missense Mutation AAA,GAA K36E NP_001019772.1

View Full Product Details